missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphopantothenate-cysteine ligase Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 468.00 €
Specifications
| Antigen | Phosphopantothenate-cysteine ligase |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Phosphopantothenate-cysteine ligase Polyclonal antibody specifically detects Phosphopantothenate-cysteine ligase in Human samples. It is validated for Western BlotSpecifications
| Phosphopantothenate-cysteine ligase | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 79717 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ11838, MGC117357, phosphopantothenoylcysteine synthetase, PPC synthetase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-311 of human PPCS (NP_078940.2). MAEMDPVAEFPQPPGAARWAEVMARFAARLGAQGRRVVLVTSGGTKVPLEARPVRFLDNFSSGRRGATSAEAFLAAGYGVLFLYRARSAFPYAHRFPPQTWLSALRPSGPALSGLLSLEAEENALPGFAEALRSYQEAAAAGTFLAVEFTTLADYLHLLQAAAQALNPLGPSAMFYLAAAVSDFYVPVSEMPEHKIQSSGGPLQITMKMVPKLLSPLVKDWAPKAFIISFKLETDPAIVINRARKALEIYQHQVVVANILESRQSFVFIVTKDSETKLLLSEEEIEKGVEIEEKIVDNLQSRHTAFIGDRN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title