missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 20% (v/v), Ricca

Product Code. 31940767
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Unit Size:
1 L
20 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940767 1 L 1 L
31940490 20 L 20 L
31940506 4 L 4 L
31940642 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31940767 Supplier Ricca Chemical Company Supplier No. R58503001A
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Phosphoric Acid
  • Available in 500 mL, 1 L, 4 L, and 20 L package sizes with natural poly and Cubitainer™ container options

Chemical Identifiers

Grade ACS

Specifications

CAS 7732-18-5, 7664-38-2
Packaging Natural Poly Container
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.