missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Ammonium Salt (1:1), TRC

Product Code. 30418017
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418017 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418017 Supplier Toronto Research Chemicals Supplier No. TRCP3591501KG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Phosphate Monobasic
CAS 7722-76-1
Quantity 1 kg
Molecular Formula H3 N . H3 O4 P
Synonym ABC, Ammonium biphosphate, Vekson AVS 25, Ammonium phosphate, Dihydrogen ammonium phosphate, Monoammonium acid phosphate, Feniks AVS 70, Furex 770
InChI Formula InChI=1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)
SMILES N.OP(=O)(O)O
IUPAC Name ammonia;phosphoric acid
Molecular Weight (g/mol) 115.026
Formula Weight 115.0034
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.