missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Diammonium Salt, TRC

Product Code. 30427710
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427710 50 g
30424363 25 g
30411019 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427710 Supplier Toronto Research Chemicals Supplier No. TRCP35920550G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-28-0
Molecular Formula 2 H3 N . H3 O4 P
Molecular Weight (g/mol) 132.06
Synonym Akoustan A, Ammonium Dibasic Phosphate, Ammonium Hydrogen Phosphate, Ammonium Hydrogen Phosphate ((NH4)2HPO4), Ammonium Monohydrogen Orthophosphate, Ammonium Monohydrogen Phosphate, Ammonium Orthophosphate Dibasic, Ammonium Phosphate ((NH4)2(HPO4)), Ammonium Phosphate Dibasic, Coaltrol LPA 445, DAP, Diammonium Acid Phosphate, Diammonium Hydrogen Orthophosphate, Diammonium Hydrogen Phosphate, Diammonium Hydrogen Phosphate ((NH4)HPO4), Diammonium Monohydrogen Phosphate, Diammonium Orthophosphate, Diammonium Phosphate, Dibasic Ammonium Phosphate, Hydrogen Diammonium Phosphate, K 2, K 2 (phosphate), Pelor, Phos-Chek 202A, Pyrozyl WAR, Secondary Ammonium Phosphate, Spartan AR 295
IUPAC Name azane;phosphoric acid
SMILES N.N.OP(=O)(O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)
Formula Weight 132.03
Chemical Name or Material Ammonium Phosphate Dibasic (AS)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.