missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, Diluted, 10% Solution, Multi-Compendial, N.F., J.T. Baker™

Product Code. 15508364 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
19 L
Packaging:
Poly Bottle
Unit Size:
19 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15508364 19 L Poly Bottle 19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508364 Supplier Avantor J.T.Baker Supplier No. 568307
  • ONLINE EXCLUSIVE PRICE
    1459.05 €
    1289.55 € / 19 L
    Save 169.50 €
    11% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Packaging Poly Bottle
Specific Gravity 1L = 1.05kg
Quantity 19 L
Formula Weight 98
Grade Multi-Compendial/NF
Chemical Name or Material Phosphoric Acid, Diluted, 10% Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.