missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Tributyl Ester, TRC

Product Code. 30134453
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134453 25 g 25g
17953075 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134453 Supplier Toronto Research Chemicals Supplier No. TRCP35944525G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-73-8
Molecular Formula C12 H27 O4 P
Molecular Weight (g/mol) 266.31
Synonym Tri-n-butyl phosphate, Phosphoric acid tributyl ester, Tributyl Phosphate, Butyl phosphate ((BuO)3PO) (6CI, 7CI), Tributyl phosphate (ACI), 941PL, Calloway 6814, Celluphos 4, Degressal SD 40, Disflamoll TB, DL 1157, DU 155, Moussex 941PL, NSC 8484, Phosflex 4, Phosphoric acid, tributyl ester, TBP, TBPA, Tri-n-butyl phosphate, Tributoxyphosphine oxide, X 8054
IUPAC Name tributyl phosphate
SMILES CCCCOP(=O)(OCCCC)OCCCC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3
Formula Weight 266.1647
Chemical Name or Material Tributylphosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.