missing translation for 'onlineSavingsMsg'
Learn More

Phosphorous acid, 98%, extra pure

Product Code. 10072760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 kg
Packaging:
Glass bottle
Unit Size:
1kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10072760 2 kg Glass bottle 2kg
10783201 500 g Glass bottle 500g
10496193 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10072760 Supplier Thermo Scientific Acros Supplier No. 201150020
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13598-36-2 | H3O3P

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13598-36-2
Molecular Formula H3O3P
MDL Number MFCD00137258
Synonym Orthophosphorous acid

Specifications

Color White
Melting Point 73.0°C
Boiling Point 200.0°C
Density 1.6500g/mL
Heavy Metals (as Pb) 20ppm max.
Packaging Glass bottle
Specific Gravity 1.65
Flash Point >100°C
Assay Percent Range 98%
Quantity 2 kg
Linear Formula H3PO3
Physical Form Crystals
Merck Index 15, 7458
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 82
Percent Purity 98%
Grade Extra Pure
Chemical Name or Material Phosphorous acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phosphorous acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.