missing translation for 'onlineSavingsMsg'
Learn More

Phosphorous acid, 98+%

Product Code. 11403494
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403494 1000 g 1000g
11413494 250 g 250g
11423494 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403494 Supplier Thermo Scientific Alfa Aesar Supplier No. A11189.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13598-36-2 | H3PO3

Phosphorus acid is used as an intermediate in the preparation of other phosphorus compounds like potassium phosphite, ammonium phosphite and calcium phosphite. It is actively involved in the preparation of phosphites like aminotris(methylenephosphonic acid) (ATMP), 1-hydroxyethane 1,1-diphosphonic acid (HEDP) and 2-phosphonobutane-1,2,4-tricarboxylic Acid (PBTC), which find application in water treatment as a scale or corrosive inhibitor. It is also used in chemical reactions as a reducing agent. Its salt, lead phosphite is used as PVC stabilizer. It is also used as precursor in the preparation of phosphine as well as an intermediate in the preparation of other phosphorus compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphorus acid is used as an intermediate in the preparation of other phosphorus compounds like potassium phosphite, ammonium phosphite and calcium phosphite. It is actively involved in the preparation of phosphites like aminotris(methylenephosphonic acid) (ATMP), 1-hydroxyethane 1,1-diphosphonic acid (HEDP) and 2-phosphonobutane-1,2,4-tricarboxylic Acid (PBTC), which find application in water treatment as a scale or corrosive inhibitor. It is also used in chemical reactions as a reducing agent. Its salt, lead phosphite is used as PVC stabilizer. It is also used as precursor in the preparation of phosphine as well as an intermediate in the preparation of other phosphorus compounds.

Solubility
Soluble in water and alcohol.

Notes
Air sensitive. Hygroscopic. Incompatible with strong bases, oxidizing agents and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13598-36-2
Molecular Formula H3PO3
MDL Number MFCD00137258
Synonym Phosphonic acid

Specifications

Melting Point 74°C
Boiling Point 200°C (decomposition)
Density 1.651
Odor Odorless
Sensitivity Hygroscopic
Quantity 1000 g
UN Number UN2834
Merck Index 14,7346
Solubility Information Soluble in water and alcohol.
Formula Weight 82
Percent Purity ≥98%
Chemical Name or Material Phosphorous acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phosphorous acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Phosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.