missing translation for 'onlineSavingsMsg'
Learn More

Phosphorous Acid, Honeywell Fluka™

Product Code. 15611980 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15611980 2 kg 2kg
15601980 100 g 100g
15621980 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15611980 Supplier Honeywell-Fluka Supplier No. 2151122KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

0.99

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13598-36-2
Molecular Formula H3O3P
MDL Number MFCD00137258

Specifications

Density 1.651g/mL at 25°C
Quantity 2 kg
UN Number 2834
Formula Weight 82g/mol
Percent Purity 99%
Chemical Name or Material Phosphorous Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.