missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus pentoxide, 98%, extra pure

Product Code. 10217483 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
250g
Product Code. Quantity Packaging unitSize
10217483 2.5 kg Glass bottle 2.5kg
10340994 250 g Glass bottle 250g
10124360 1 kg Glass bottle 1kg
11994091 10 kg Plastic drum 10kg
4 options
This item is not returnable. View return policy

Product Code. 10217483

Brand: Thermo Scientific Acros 200890025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-56-3 | O5P2 | 141.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
Synonym Phosphoric anhydride
IUPAC Name tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

Specifications

Melting Point 340.0°C to 360.0°C
Color White
Physical Form Hygroscopic Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 98%
Linear Formula P2O5
Fieser 01,871; 07,291; 10,319; 14,151; 15,267
Merck Index 15,7467
Solubility Information Solubility in water: 85% (20°C); reacts forming h3po4 with evol.of heat
Formula Weight 141.94
Percent Purity 98%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Phosphorus pentoxide
Show More Show Less
compliance-icons
Product Identifier
  • Phosphorus pentoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Phosphorus oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.