missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) oxide, Puratronic™, 99.9925% (metals basis), Thermo Scientific Chemicals

Product Code. 11396957
Change view
Click to view available options
Quantity:
10 g
100 g
5 x 100 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396957 5 x 100 g 500g
11386957 10 g 10g
11306967 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396957 Supplier Thermo Scientific Alfa Aesar Supplier No. 010827.59
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A dehydrating agent often used as a desiccant for bacterial cultural extracts analyzed by gas-liquid chromatography. Used for the preparation of phosphoric acid, phosphorus compound, it can also be used for the preparation of high-purity phosphoric acid. Also used in sugar refining and a catalyst for air blowing of asphalt, reagent in the efficient and modified Biginelli-type synthesis of 3,4-dihydro-1H-indeno[1,2-d]pyrimidine-2,5-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
IUPAC Name tricyclo[3.3.1.1⊃3,⊃7]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

Specifications

Boiling Point 360°C (sublimation)
Melting Point 340°C
Physical Form Powder
Quantity 5 x 100 g
UN Number UN1807
Merck Index 14,7355
Solubility Information Soluble in sulfuric acid. Insoluble in acetone and ammonia.
Formula Weight 141.94
Percent Purity 99.9925%
Grade Puratronic
Odor Weak
Sensitivity Moisture sensitive
Density 2.39 g/mL
Assay (metals basis)
Chemical Name or Material Phosphorus(V) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.