missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phosphorylating Phosphoramidite, standard grade, 20-400 finish bottle

Product Code. 17917182 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μmol
Unit Size:
100µmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17917182 100 μmol 100µmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17917182 Supplier Thermo Scientific™ Supplier No. 27179409

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phosphorylating phosphoramidite can be utilized to achieve phosphorylation of the 5' oligo terminus (also known as Phosphate-ON or Chemical Phosphorylation Reagent (CPR)).

Phosphorylating phosphoramidite can be utilized to achieve phosphorylation of the 5' oligo terminus (also known as Phosphate-ON or Chemical Phosphorylation Reagent (CPR)). Chemical phosphorylation is a cost-effective substitute to enzymatic methods (via T4 kinase and ATP), allowing efficient introduction of terminal phosphate groups. Oligos with a 5'-phosphate are conducive to a number of various applications utilizing the ligation of oligos to each other, such as NGS adapters, cloning, and gene construction. This product is supplied in a 30 mL amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 2-[2-(4,4-Dimethoxytrityloxy)ethylsulfonyl]ethyl-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C34H45F3N2O7PS
Formula weight: 656.66 g/mol
HPLC Purity: ≥95.0%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Expedite Synthesizers
Product Type Phosphorylating CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type None (Modifier)
Shipping Condition Room Temperature
Purity 95%
Quantity 100 μmol
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type None (Modifier)
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.