missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ PhosVer™ 3 Phosphate Reagent Powder Pillows
Click to view available options
Quantity:
10 mL
Unit Size:
Pack of 100
Specifications
Specifications
| Format | Solid |
| Quantity | 10 mL |
Safety and Handling
Product Identifier
- PhosVer3 Phosphate Reagent
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P363-Wash contaminated clothing before reuse.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Potassium disulfate,Sodium molybdate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction