missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Photo Check Solution

Product Code. 10430673
Change view
Click to view available options
For Use With (Equipment):
Photometers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10430673 Photometers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10430673 Supplier WTW™ Supplier No. 250497
  • ONLINE EXCLUSIVE PRICE
    188.00 €
    180.00 € / Pack of 1
    Save 8.00 €
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-made reagent presentations for the digestion of nitrate for the determination of the total nitrogen as well as heavy metals contain all required reagents for thermal digestion

Thermal digestions are required in water analysis for the total determination of materials as well as heavy metals. Ready-made reagent presentations for the digestion of nitrate for the determination of the total nitrogen as well as heavy metals contain all required reagents for thermal digestion. These ready-made cuvettes or the simple preparation in empty cuvettes with covers makes this process safe, easy and quick. Then, the prepared samples in 16 mm cuvettes are digested in the thermoreactor with selectable standard programs. Ready-made test kits are available for the determination of COD, total nitrogen and total phosphorus, which already contain the digestion reagent and digested and measured in one and the same cuvette.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Photo Check Solutions
For Use With (Equipment) Photometers
compliance-icons compliance-icons
Product Identifier
  • Crack-Set 20
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Potassium persulphaye, Potassium carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.