missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pHrodo™ Dyes for Amine Labeling

Product Code. 15913783
Change view
Click to view available options
Color:
Deep Red
Green
Red
Excitation/Emission:
505/525
560/585
640/655
Reactive Moiety:
Active Ester, STP Ester
TFP ester
Quantity:
1 mg
3 x 100 μg
Unit Size:
100g
1mg
300µg
Pack of 3
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Excitation/Emission Reactive Moiety Quantity Unit Size
15913783 Green 505/525 Active Ester, STP Ester 1 mg 1mg
15875872 Red 560/585 Active Ester, STP Ester 3 x 100 μg Pack of 3
17171919 Red 560/585 Active Ester, STP Ester 3 x 100 μg 100g
15903783 Red 560/585 Active Ester, STP Ester 1 mg 1mg
15700847 Green 505/525 Active Ester, STP Ester 3 x 100 μg Pack of 3
17158292 Deep Red 640/655 TFP ester 3 x 100 μg 300µg
17168292 Deep Red 640/655 TFP ester 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15913783 Supplier Invitrogen™ Supplier No. P36012
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Amine-reactive and TFP ester pHrodo dyes enable accurate measurement of cell endocytosis and phagocytosis, with low signal-to-background, for use in live cell assays.

Perform cell internalization studies with low signal-to-background and high specificity using the pHrodo pH sensor dyes, including pHrodo Red, Green, and Deep Red dyes. These dyes can be conjugated to biomolecules such as antibodies, and become highly fluorescent only when found in acidic environments, such as those of cell lysosomes and endosomes. This enables detection of the conjugated biomolecule in the live cell assay without the need for wash steps or quencher dyes in the experimental workflow.

pHrodo Red, Green, and Deep Red dyes
The amine-reactive pH-sensitive pHrodo Red, Green, and Deep Red dyes are optimized for the creation of bioconjugates used in the study of antibody internalization, endocytosis, and phagocytosis in live cell assays. Amine-reactive pHrodo dyes enable you to label primary amines on an antibody, protein, cell, virus, or other amine-containing biomolecule to create a conjugate that fluoresces inside acidic environments but shows little to no signal at neutral pH. The lack of fluorescence of pHrodo dyes in a typical extracellular environment eliminates the need for wash steps or quencher dyes in the experimental workflow.

pHrodo iFL Red and Green STP Ester dyes
pHrodo iFL Red and Green STP Ester dyes are improved versions of the original pHrodo Red and Green dyes and are optimized to create more soluble conjugates than the original dyes. This enhanced property makes them useful for the labeling of antibodies that may otherwise precipitate out of solution during conjugation.

pHrodo Deep Red TFP Ester
The pHrodo Deep Red TFP Ester is distinct from pHrodo iFL Red and Green Ester dyes in that it has a lower pKa and, as a result, shows both improved signal-to-background ratios and fluoresces later in the endocytic pathway. Our low-background pHrodo Deep Red dye makes it easier than ever to study internalization with a higher certainty of results and less optimization because pHrodo Deep Red dye only fluoresces in the late endosome and lysosome.

Features and benefits of the pHrodo dyes for amine labeling

• Customization—use pH-sensitive pHrodo dyes to make pH-sensitive bioconjugates of your choice
• Speed and accuracy—get faster and more accurate results than with any other endocytosis or phagocytosis assay, no need for wash steps or quenchers
• Multiplexing—multiplex with green or red fluorescent markers such as GFP or RFP with three different color choices
• Superior signal window—with pHrodo Deep Red dye’s minimal background at neutral pH, maximal signal-to-noise ratio is enabled for certainty of results
• Low pKa—pHrodo Deep Red dye eliminates non-specific signal from non-internalized cargo, resulting in fluorescence only in late endosomes and lysosomes

The increase in fluorescence of pHrodo dyes as the pH acidifies correlates with the acidification of intracellular vesicles, making these dyes valuable tools for the study of endocytosis or phagocytosis and their regulation by environmental factors, drugs, or pathogens. The spectral properties of pHrodo dyes also make them useful for multi-color experiments. pHrodo iFl Red, iFL Green, and Deep Red dyes can be detected using most standard red, green, and deep red fluorescent filter sets. pHrodo dye conjugates have been validated for use in a variety of applications, including flow cytometry, fluorescent microscopy, and high content screening (HCS).

pHrodo Deep Red dye has a lower pKa by design and shows no signal in neutral conditions, fluorescing only in acidic environments. This unique property enables rapid assay development and certainty of results when investigating antibody internalization and endocytic/phagocytic pathways. pHrodo Deep Red dye enables better discrimination of internalized cargo from outside the cell, whereas pHrodo iFl Red and Green dyes fluoresce in the early endosome.

Order Info

Store in freezer (-5 to -30°C) and protect from light.

TRUSTED_SUSTAINABILITY

Specifications

Description pHrodo iFL Green STP ester, amine reactive dye
Quantity 1 mg
Molecular Weight (g/mol) ∼1000
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, STP Ester
Label or Dye pHrodo™ Green
Label Type pHrodo (pH sensitive)
Color Green
Product Line pHrodo
Excitation/Emission 505/525
Show More Show Less
Is there an alternative chaotrope for 8M guanidine-HCl solution used for determining degree of labeling (DOL) for pHrodo Dyes for Amine Labeling (Cat. Nos. P36010, P36011, P36012, P36013)?

Yes. A solution of 8 M urea in 20 mM HEPES can be used as an alternative for 8 M guanidine-HCl solution in the degree of labeling (DOL) determination protocol for pHrodo Dyes for Amine Labeling (Cat. Nos. P36010, P36011, P36012, P36013).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.