missing translation for 'onlineSavingsMsg'
Learn More

Phthalazine, 98%

Product Code. 10061360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
10g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10061360 10g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10061360 Supplier Thermo Scientific Acros Supplier No. 131050100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 253-52-1 | C8H6N2 | 130.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 253-52-1
Melting Point 88.0°C to 92.0°C
Color Beige to Brown
Boiling Point 315.0°C to 317.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H6N2
MDL Number MFCD00006908
Quantity 10 g
Beilstein 23, 174
Merck Index 15, 7482
Synonym 2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,methanol,benzene,ethyl aceta,te; less soluble in ehter; insoluble in ligroine
InChI Key LFSXCDWNBUNEEM-UHFFFAOYSA-N
SMILES C1=CC=C2C=NN=CC2=C1
IUPAC Name phthalazine
Molecular Weight (g/mol) 130.15
PubChem CID 9207
ChEBI CHEBI:36597
Formula Weight 130.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Phthalazine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.