missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Acid, TRC

Product Code. 30425428
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425428 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425428 Supplier Toronto Research Chemicals Supplier No. TRCP384480100G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic Acid
CAS 88-99-3
Molecular Formula C8 H6 O4
Quantity 100 g
Synonym 1,2-Benzenedicarboxylic acid, Phthalic acid (8CI), M 2, NSC 5348, Sunftal 20, o-Benzenedicarboxylic acid, o-Carboxybenzoic acid, o-Dicarboxybenzene, Benzene-1,2-dicarboxylic acid, Phthalic acid
InChI Formula InChI=1S/C8H6O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H,9,10)(H,11,12)
SMILES OC(=O)c1ccccc1C(=O)O
IUPAC Name phthalic acid
Molecular Weight (g/mol) 166.1308
Formula Weight 166.0266
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.