missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Anhydride, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15557804 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15557804 2.5 kg Poly Bottle 2.5kg
15547804 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15557804 Supplier Avantor J.T.Baker Supplier No. 027205

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 85-44-9
Molecular Formula C8H4O3
Molecular Weight (g/mol) 148.12
MDL Number MFCD00005918
InChI Key LGRFSURHDFAFJT-UHFFFAOYSA-N
Synonym phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen
PubChem CID 6811
ChEBI CHEBI:36605
SMILES O=C1OC(=O)C2=CC=CC=C12

Specifications

Flash Point 152.00°C
Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1L = 1.53kg
IUPAC Name 2-benzofuran-1,3-dione
Formula Weight 148.12
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Phthalic Anhydride
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.