missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Anhydride (May Contain Up To 15% Of Phthalic Acid), TRC

Product Code. 30411131
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411131 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411131 Supplier Toronto Research Chemicals Supplier No. TRCP38447350G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic anhydride
CAS 85-44-9
Molecular Formula C8 H4 O3
Quantity 50 g
Synonym 1,3-Isobenzofurandione, Phthalic anhydride (8CI), 1,2-Benzenedicarboxylic anhydride, 1,3-Phthalandione, 2-Benzofuran-1,3-dione, ADP 1200, Araldite HT 901, ESEN, HT 901, NSC 10431, OP 392, Phthalandione, Phthalanhydride, Phthalic acid anhydride, Retarder AK, Retarder B-C, Retarder ESEN, Retarder PD, Rikacid PA, Sconoc 5, Sconoc 7, TGL 6525, Vulkalent B/C
InChI Formula InChI=1S/C8H4O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-4H
SMILES O=C1OC(=O)c2ccccc12
IUPAC Name 2-benzofuran-1,3-dione
Molecular Weight (g/mol) 148.12
Formula Weight 148.016
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.