missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Hydrazide, TRC

Product Code. 30428380
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428380 10 g -
30427709 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30428380 Supplier Toronto Research Chemicals Supplier No. TRCP38451010G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-69-8
Molecular Formula C8 H6 N2 O2
Molecular Weight (g/mol) 162.15
Synonym 2,3-Dihydro-1,4-phthalazinedione, 1,2-Benzenedicarboxylic acid, cyclic hydrazide, 1,4-Dihydroxyphthalazine, 1,4-Phthalazinediol, 4-Hydroxyphthalazin-1(2H)-one, Hydrazine, 1,2-(1,2-phenylenedicarbonyl)-, N,N'-Phthaloylhydrazine, NSC 201511, NSC 651, Phthalazine-1,4(2H,3H)-dione, Phthalhydrazide, Phthalic acid cyclic hydrazide, Phthalocyclohydrazide
IUPAC Name 2,3-dihydrophthalazine-1,4-dione
SMILES O=C1NNC(=O)c2ccccc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H6N2O2/c11-7-5-3-1-2-4-6(5)8(12)10-9-7/h1-4H,(H,9,11)(H,10,12)
Formula Weight 162.0429
Chemical Name or Material Phthalic Hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.