missing translation for 'onlineSavingsMsg'
Learn More

Phthalimidoacetaldehyde diethyl acetal, 99%

Product Code. 11418407
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418407 25 g 25g
11408407 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418407 Supplier Thermo Scientific Alfa Aesar Supplier No. A18491.14
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78902-09-7 | C14H17NO4 | 263.29 g/mol

Phthalimidoacetaldehyde diethyl acetal is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalimidoacetaldehyde diethyl acetal is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78902-09-7
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.29
MDL Number MFCD00005901
InChI Key GEFXJJJQUSEHLV-UHFFFAOYSA-N
Synonym phthalimidoacetaldehyde diethyl acetal, 2-2,2-diethoxyethyl isoindoline-1,3-dione, n-2,2-diethoxyethyl phthalimide, 2-phthalimidoacetaldehyde diethyl acetal, 2-phthalimido acetaldehyde diethylacetal, 2-2,2-diethoxyethyl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl, 2-2,2-diethoxyethyl-1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl benzo c azoline-1,3-dione, acmc-209pfw
PubChem CID 315286
IUPAC Name 2-(2,2-diethoxyethyl)isoindole-1,3-dione
SMILES CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC

Specifications

Melting Point 71°C to 74°C
Quantity 25 g
Beilstein 215793
Solubility Information Insoluble in water.
Formula Weight 263.3
Percent Purity 99%
Chemical Name or Material Phthalimidoacetaldehyde diethyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.