missing translation for 'onlineSavingsMsg'
Learn More

Phthalocyanine, 93%, BLD Pharm™

Product Code. 31109932
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31109932 25 g
31095585 5 g
31097234 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31109932 Supplier BLD Pharm Supplier No. BD14793425g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 574-93-6
Molecular Formula C32H18N8
MDL Number MFCD00005085
Quantity 25 g
SMILES C1(C2=N3)=C(C=CC=C1)C(N2)=NC4=NC(C5=CC=CC=C54)=NC(N6)=C7C=CC=CC7=C6N=C8C9=CC=CC=C9C3=N8
Molecular Weight (g/mol) 514.54
Percent Purity 93%
Chemical Name or Material Phthalocyanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.