missing translation for 'onlineSavingsMsg'
Learn More

Phthaloyl chloride, 90%, AcroSeal™, Thermo Scientific™

Product Code. 13505000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13505000 100 mL AcroSeal™ Glass bottle 100mL
13525000 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13505000 Supplier Thermo Scientific Acros Supplier No. 417971000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-95-9
InChI Key FYXKZNLBZKRYSS-UHFFFAOYSA-N
Synonym phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride
PubChem CID 6955
IUPAC Name benzene-1,2-dicarbonyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl

Specifications

Melting Point 12.0°C
Density 1.4000g/mL
Boiling Point 269.0°C to 271.0°C
Flash Point 113°C
Assay Percent Range 90%
Packaging AcroSeal™ Glass bottle
Molecular Formula C8H4Cl2O2
Linear Formula C6H4(COCl)2
Refractive Index 1.5674 to 1.5694
Quantity 100 mL
Specific Gravity 1.4
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether, decomposes in alcohol
Molecular Weight (g/mol) 203.018
Formula Weight 203.02
Percent Purity 90%
Chemical Name or Material Phthaloyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.