missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Blood Direct PCR Master Mix

Product Code. 15280114 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
15280114 100 Reactions 100 reactions
15290114 500 Reactions 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15280114 Supplier Thermo Scientific™ Supplier No. F175S
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific Phusion Blood Direct PCR Master Mix is designed for amplification of DNA from whole blood. It eliminates the need for a separate DNA purification step prior to PCR.

Thermo Scientific Phusion Blood Direct PCR Master Mix is designed for amplification of DNA from whole blood. It eliminates the need for a separate DNA purification step prior to PCR. The modified Phusion Hot Start II High-Fidelity DNA Polymerase is resistant to main PCR inhibitors present in blood and retains its activity at blood concentrations of up to 40% in the PCR reaction.

This accurate proofreading DNA polymerase makes the Phusion Blood Direct PCR Master Mix an ideal choice for demanding applications, such as cloning. Phusion Blood Direct PCR Master Mix also allows analysis of blood samples with different preservatives (e.g., heparin, EDTA, citrate), storage conditions, and from various species.

Highlights

  • Direct PCR allows sample to be added directly to PCR reaction, reducing time-consuming and expensive DNA purification steps
  • Available Dilution & Storage protocol allows several PCR reactions from one sample
  • Specially engineered Phire Hot Start II DNA polymerase for extremely short PCR protocol times
  • Master mix format with premixed gel loading dye minimizes risk of cross-contamination, reduces sample handling, and allows direct loading on gel

Applications

  • Genotyping
  • Transgene detection
  • Knockout analysis
  • Sequencing
  • Cloning

Notes

  • Annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • 2X Phusion Blood Direct PCR Master Mix, 1 mL
• Universal Control Primer Mix, 40 μL
• 50 mM EDTA, 0.5 mL
• 50 mM DMSO, 0.5 mL
• 50 mM MgCl2, 1.5 mL
• Nuclease-free water, 1.25 mL
• O'GeneRuler Express DNA Ladder, 50 μg (sufficient for 100 applications)

Store at -20°C.
GC-Rich PCR Performance High
PCR Method Direct PCR
Polymerase Phusion Blood II DNA Polymerase
Reaction Speed Fast
Includes Label or Dye Yes
Product Type Direct PCR Master Mix
Quantity 100 reactions
Shipping Condition Wet or Dry Ice
For Use With (Application) Genotyping
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Sample Type Blood
Starting Material DNA
Show More Show Less
What is your recommendation to take a sample from plant or animal tissue for Direct PCR?

In order to obtain best results from Direct PCR, it is important to use a very small amount of starting materials. The recommended size is 0.3-0.5 mm in diameter. We recommend using Tissue Puncher 0.3 mm (Cat. No. F-200S), which is a convenient tool for sampling and is suitable for both plant and animal tissues.

Why are Control primers provided in the Direct PCR Kits?

It is very important to include a control reaction in the Direct PCR to ensure that the PCR conditions are optimal for the specific tissue or plant material used. Control primers are universal primers that amplify a small fragment from a conserved genome region. If the reaction with control primers fails, the sample material may not be suitable for Direct PCR, or PCR conditions need optimization.

Can I use plant or tissue sample which is not on the list of the Direct PCR Master Mix and Kit tested samples?

You can test any plant or tissue sample. However, it is recommended to first optimize the reaction. When working with new sample materials, start with the Dilution & Storage protocol as it allows several PCR reactions to be performed from the same sample during optimization. It is recommended to perform a positive control with purified sample DNA to ensure that the PCR conditions are working. If the positive control with purified DNA fails, the PCR conditions should be optimized before continuing further.

Can you amplify the same sample materials with the Direct PCR Master Mixes as with Direct PCR Kits?

Yes, the Direct PCR Master Mixes are upgrades of the Direct PCR kits and thus are suitable for the same sample materials.

What is the highest blood concentration possible in the reaction when using Phusion Blood Direct PCR Master Mix and Kit?

We recommend using blood concentration up to 20%; however, we had successful amplifications with up to 40% of blood in the reaction mix. In some cases optimization may be required to achieve this.

Why is EDTA included in the Phusion Blood Direct PCR Master Mix and Kits?

Extensive testing has shown 3.0 mM MgCl2 to be optimal concentration for most PCR reactions using whole blood. There is a separate tube of 50 mM MgCl2 (F-510MG) included in the kit for adjusting the MgCl2 concentration (up to 4.5 mM if necessary) for reactions containing very high amounts of blood. It should be noted though that excess MgCl2 may result in spurious PCR products. If unspecific products are created with the default reaction buffer (providing 3.0 mM MgCl2), the effective MgCl2 concentration can be decreased by adding the chelating agent EDTA, which is included in the kit. Typically, adding 0.5 to 1.0 µL of 50 mM EDTA to a 20 µL reaction is sufficient to eliminate non-specific products. Note that the optimal conditions will depend on the primers, the percentage of blood in the reaction and/or the type of card used, since anticoagulants and other chemicals impregnating the cards can alter the available Mg2+ concentration.

Do the Direct PCR Master Mixes and Kits come with tissue punchers?

No, tissue punchers are not provided. You can purchase 0.3 mm Tissue Punchers (Cat. No. F-200S), or cut the sample with a scalpel. Some scientists use 100 µL pipette tips to take a sample.

Do the Direct PCR Master Mixes work in Dilution & Storage protocol?

Yes, similar to the Direct PCR Kits, Direct PCR Master Mixes work in both Direct and Dilution & Storage protocols. Direct PCR from blood is performed with Direct protocol only.

What if I want to clone or sequence the PCR product from a Direct PCR kit? Can I use the kit with Phusion instead of Phire?

Direct PCR Kits and Master Mixes employ Phire Hot Start II DNA Polymerase or Phusion Hot Start II Polymerase (depending on the kit), that both possess the following activities: 5'-3' DNA polymerase activity and a weak 3'-5' exonuclease activity. When cloning fragments amplified with these DNA Polymerases, blunt end cloning is recommended. If TA cloning is required, it can be performed by adding A overhangs to the blunt PCR product. However, before adding the overhangs, it is very important to remove all the DNA polymerase by purifying the PCR product carefully. Any remaining Phusion or Phire DNA Polymerases will degrade the A overhangs, creating blunt ends again. All Direct PCR kits are compatible with sequencing techniques. The green dye present in the Direct Master Mix kits does not interfere with the sequencing reactions.

Does Phusion Blood Direct PCR Kit and Master Mix have an IVD-CE mark?

No.

Can I use Phusion Blood Direct PCR Kit/Master Mix for allele-specific PCR directly from blood (e.g. HLA-typing)?

Phusion Blood DNA Polymerase has a strong 3'- 5' exonuclease activity which is able to cleave the mismatched base from the 3' end and continue amplification. If the mismatch is located at the 3' end and is not protected from cleavage by any modification, Phusion polymerase will cut the mismatched base.

After performing Direct PCR, how should I store the PCR products amplified directly from the sample?

After PCR, it is recommended to centrifuge the PCR reactions at 1000 x g for 1-3 minutes to collect the supernatant for subsequent analysis. We recommend storing the supernatant at -20 degrees C

Are there any restrictions to the size of the amplicon in Direct PCR?

The optimal length for Direct PCR from liquid samples is up to 5 kb and from solid samples up to 1 kb. However, the maximum length depends on the sample type, sample size and the reaction volume. For example we have amplified a 7.5 kb fragment directly from human blood and tissue samples, and a 3 kb fragment from mouse ear and 5 kb fragment from plant leaves.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.