missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Flash High-Fidelity PCR Master Mix

Product Code. p-4526973 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10043967 100 Reactions 100 reactions
10659009 500 Reactions 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 10043967 Supplier Thermo Scientific™ Supplier No. F548S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Thermo Scientific Phusion Flash PCR Master Mix was developed to save valuable laboratory time.

Thermo Scientific Phusion Flash PCR Master Mix was developed to save valuable laboratory time. Ready-to-use 2X master mix preserves the fidelity and the yield in reactions when using very short PCR protocols. The unique master mix composition permits usage of short cycling protocols with both low and high complexity DNA templates (15 seconds per kilobase or less). The master mix utilizes Phusion Flash II DNA Polymerase, a modified proofreading DNA polymerase derived from Phusion Hot Start II High-Fidelity DNA Polymerase.

Features

  • Extreme speed with extension times of 15 s/kb or less
  • Hot start modification allows 'zero-time reactivation'
  • Accurate proofreading DNA polymerase with a fidelity of 25X Taq
  • High yields in reduced cycling time
  • Minimized pipetting steps

Applications

  • Fast PCR
  • High-fidelity PCR
  • Difficult (GC-rich) templates
  • Template generation for sequencing Multiplex PCR
  • Long-range PCR
  • Cloning & mutagenesis
  • Microarray

Notes

  • Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Phusion Flash High-Fidelity PCR Master Mix (1 mL)

Store at -20°C.
GC-Rich PCR Performance High
Polymerase Phusion Flash II DNA Polymerase
Reaction Speed Fast
Product Type High-Fidelity PCR Master Mix
Quantity 100 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) 25X
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 20 kb or less
Color Colorless
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.