missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion™ Hot Start II DNA Polymerase & dNTP Mix (10 mM each)

Product Code. 17174519 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2000 reactions
Unit Size:
2000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17174519 2000 reactions 2000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 17174519 Supplier Thermo Scientific™ Supplier No. F549N

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

This package includes Phusion Hot Start II High-Fidelity DNA Polymerase (4 x 500 U) and high quality dNTP Mix (10 mM each, 2 x 1 mL).

Thermo Scientific Phusion Hot Start II High-Fidelity DNA Polymerase sets a high standard for high performance PCR. Featuring an error rate 52-fold lower than that of Taq and 6-fold lower than that of Pfu, this polymerase is an excellent choice for cloning and other applications that require high fidelity and offers robust performance with short protocol times, even in the presence of PCR inhibitors. Phusion Hot Start II High-Fidelity DNA Polymerase generates higher yields with lower enzyme amounts than other DNA polymerases.

With Phusion Hot Start II High-Fidelity DNA Polymerase, amplification proceeds without the production of nonspecific products due to the combination of Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand that inhibits DNA polymerase activity at room temperature. The Affibody ligand also inhibits the 3' to 5' exonuclease activity of the polymerase, thus preventing degradation of primers and template DNA during reaction setup. At temperatures that promote polymerase activity, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II High-Fidelity DNA Polymerase is immediately reactivated at high temperatures and does not require a separate activation step in PCR protocols.

Features

  • High fidelity (52X Taq)
  • No non-specific amplification and primer degradation during reaction setup
  • Fast PCR due to short extension times (15–30 s/kb)
  • Increased product yields with minimal enzyme amounts

Applications

  • Standard PCR
  • qPCR
  • High fidelity and long PCR
  • LAMP-PCR
  • cDNA synthesis
  • RT-PCR, RDA, MDA
  • DNA labeling and sequencing

dNTP Mix

Thermo Scientific dNTP Mix (Cat. No. R0192) contains pre-mixed aqueous solutions of dATP, dCTP, dGTP, and dTTP, each at a final concentration of 10 mM. The nucleotides have greater than 99% purity and are free of nuclease activities, as well as human and E. coli DNA. Mixes offer the possibility of reducing the number of pipetting steps and the risk of reaction setup errors.

  • Greater than 99% purity confirmed by HPLC
  • Free of human and E. coli DNA
  • Stable after multiple freeze-thaw cycles
  • Up to 95% of dNTPs remain in triphosphate form after 7 weeks at room temperature
  • Up to 90% of dNTPs remain in triphosphate form after 30 cycles of PCR (1 minute at 94°C; 3 minutes at 72°C)

Notes

  • This package includes Phusion Hot Start II High-Fidelity DNA Polymerase (4 x 500 U) and high-quality dNTP Mix (10 mM each, 2 x 1 mL).
  • Annealing rules for Phusion DNA polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Content And Storage • Phusion Hot Start II High-Fidelity DNA Polymerase (4 x 500 U at 2 U/μL)
• 10 mM dNTP Mix (2 x 1 mL)
• 5X Phusion HF & GC Buffers
• DMSO
• 50 mM MgCl2 solution

Store at –5°C to –30°C.
Detection Method Primer-probe
GC-Rich PCR Performance High
Polymerase Phusion Hot Start II DNA Polymerase
Reaction Speed Fast
Product Type Hot Start DNA Polymerase and dNTP Mix
Quantity 2000 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) 52X
Hot Start Built-In Hot Start
No. of Reactions 2000 Reactions
Overhang Blunt
Reaction Format Separate Components
Size (Final Product) 20 kb or less
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.