Learn More
Descrizione
Thermo Scientific Phusion U DNA Polymerase is a novel engineered high fidelity enzyme developed using fusion technology. Due to a proprietary mutation in the so called dUTP binding pocket of Phusion polymerase, Phusion U polymerase overcomes an important limitation of proofreading enzymes: it is able to incorporate dUTP and read through uracil present in DNA templates.
In addition to the uracil usage possibility, Phusion U polymerase features all the superior properties of other Phusion DNA polymerases: great accuracy, speed, ability to amplify long amplicons up to 20 kb, and a high specificity with Affibody-based hot start. These features make Phusion U Hot Start DNA Polymerase an excellent choice for such important applications as amplification of bisulphite-converted or damaged DNA as well as use of carryover contamination control.
Phusion U Hot Start Green DNA Polymerase is a combination of Phusion U Hot Start DNA Polymerase and 5X Phusion Green buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with Phusion U polymerase performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.
Highlights
- Accuracy—high fidelity DNA polymerase (25X Taq)
- Uracil-tolerance—engineered to incorporate dUTP and amplify uracil containing templates
- Specificity—hot start for reduced nonspecific amplification and primer degradation
- Speed—short extension times (15–30 s/kb)
- Green format—permits direct loading of PCR products on gels
Applications
- Amplification of bisulphite-converted DNA
- Amplification of damaged or aged DNA
- Carry-over contamination control
- Uracil-excision based (USER) cloning methods
Using Phusion DNA polymerases
Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).
Specifica
Specifica
| Concentration | 2 U/μL |
| Content And Storage | • Phusion U Hot Start DNA Polymerase, 2 U/μL • 5X Phusion HF & GC Buffers • DMSO • 50 mM MgCl2 solution Phusion HF Buffer and Phusion GC Buffers each provide 1.5 mM MgCl2 in the final 1X concentration. Store at -20°C. |
| GC-Rich PCR Performance | High |
| Polymerase | Phusion U Hot Start DNA Polymerase |
| Reaction Speed | Fast |
| Product Type | Hot Start DNA Polymerase |
| Quantity | 100 units |
| Shipping Condition | Dry Ice |
| For Use With (Application) | Hot-start PCR, High-fidelity PCR |
| Fidelity (vs. Taq) | 25X |
| Vedi altri risultati |
For Research Use Only. Not for use in diagnostic procedures.
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.