Learn More
uvex Phynomic AirLite A ESD Safety Glove
Product Code. p-7228497 Shop All Uvex Products
Click to view available options
Size:
5
6
7
8
9
10
11
12
Unit Size:
10 pairs
Product Code. 17767569
Brand: uvex 6003805
Only null left
Add to basket
Add to basket
The Uvex phynomic airLite ESD is a light safety glove that offers a noticeable difference in wearer comfort.
  • The lightest and most sensitive uvex phynomic cut protection glove in Cut Level A
  • Outer material made from polyamide, elastane and carbon
  • Breathable 'airLite aqua-polymer coating on palm and fingertips
  • Safety glove with touchscreen function – for use on almost all screens, tablets and mobile phones
  • Safety glove with ESD function in line with DIN EN 16350:2014 – protects workpiece against electrostatic charge
  • Good grip in dry and slightly damp areas
  • Free from accelerators, and health protection and skin compatibility dermatologically approved (proDERM)
  • Very lightweight and breathable safety glove
  • Outstanding sensitivity thanks to new breathable 'airLite' aqua-polymer coating combined with fine liner (18 gauge)
  • Safety gloves with knitted cuff and ergonomic fit
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 388 (3 1 1 0 X) and DIN EN 16350:2014
Coating Material Aqua-Polymer Foam
Color Black
Disposable Reusable
Exterior Finish Smooth
Hand Ambidextrous
Laboratory/Cleanroom Use Not tested
Latex Free Yes
Material Polyamide, Elastane, Carbon Fiber
Powdered Powder Free
Product Type ESD Gloves
Size 5
Sterility Non-sterile