Learn More
uvex Phynomic AirLite B ESD Safety Glove
Product Code. 17707599 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17707599 6 10 pairs
17717599 7 10 pairs
17727599 8 10 pairs
17737599 9 10 pairs
17747599 10 10 pairs
17757599 11 10 pairs
17767599 12 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17707599 Supplier uvex Supplier No. 6007806
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
The Uvex phynomic airLite ESD is a light safety glove that offers a noticeable difference in wearer comfort.
  • The lightest and most sensitive uvex phynomic cut protection glove in Cut Level B
  • Outer material made from innovative Dyneema Diamond 2.0 cut protection fibers, polyamide, elastane and carbon
  • Breathable 'airLite' aqua-polymer coating on palm and fingertips
  • Safety glove free from glass and steel fibers
  • Safety glove with touchscreen function – for use on almost all screens, tablets and mobile phones
  • Safety glove with ESD function in line with DIN EN 16350:2014 – protects workpiece against electrostatic charge
  • Good cut protection (Level B)
  • Good grip in dry and slightly damp areas
  • Free from accelerators, and health protection and skin compatibility dermatologically approved (proDERM)
  • Very lightweight and breathable safety glove
  • Outstanding sensitivity thanks to new breathable 'airLite' aqua-polymer coating combined with fine liner (18 gauge)
  • Safety gloves with knitted cuff and ergonomic fit
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 388 (3 X 3 2 B) and DIN EN 16350:2014
Coating Material Aqua-Polymer Foam
Color Light Blue, Black
Disposable Reusable
Exterior Finish Smooth
Hand Ambidextrous
Laboratory/Cleanroom Use Not tested
Latex Free Yes
Material Polyamide, Elastane, Carbon Fiber
Powdered Powder Free
Product Type ESD Gloves
Size 6
Sterility Non-sterile