missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic All-Round Safety Gloves

Product Code. 17210361 Shop All Uvex Products
Change view
Click to view available options
Size:
5
6
7
8
9
10
11
12
Unit Size:
10 pairs
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17210361 5 10 pairs
17220361 6 10 pairs
17230361 7 10 pairs
17240361 8 10 pairs
17250361 9 10 pairs
17260361 10 10 pairs
17270361 11 10 pairs
17280361 12 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17210361 Supplier uvex Supplier No. 6004905
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight, dirt-resistant gloves are constructed from a blend of polyamide and elastane, with aqua-polymer foam coating on the palm and fingertips to repel moisture.

  • Lightweight construction acts as a second-skin to maintain a high level of feeling
  • Aqua-polymer foam coating is moisture repellent and flexible
  • Breathable materials help prevent excessive perspiration
  • Dirt-resistant exterior is ideal for mechanical tasks
  • Enhanced grip in dry and slightly damp applications
  • 3D Ergo technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Gray, Black
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Polyamide, Elastane
Product Type Mechanical Protection Gloves
Size 5
EN388 Standard 3131X
For Use With (Application) Assembly, Maintanence, Shipping/Logistics, Precision Work, and Repairs
Gender Unisex
Glove Cuff Style Knit
Series Phynomic All-Round
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.