missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic C5 Cut Protection Gloves
GREENER_CHOICE

Product Code. 17250391 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17250391 7 10 pairs
17240391 6 10 pairs
17260391 8 10 pairs
17270391 9 10 pairs
17280391 10 10 pairs
17290391 11 10 pairs
17200401 12 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17250391 Proveedor uvex N.º de proveedor 6008107
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones

Lightweight, sensitive gloves provide level C cut resistance for mechanical activities. Aqua-polymer foam coating is breathable and enhances grip capabilities in dry or slightly damp applications.

  • Lightweight construction acts as a second-skin for tactile sensitivity
  • Optimum material composition provides level C cut protection
  • Aqua-polymer foam coating is breathable and repels dampness
  • Gloves are made from materials that prevent excessive perspiration
  • Enhanced grip in dry, and slightly damp applications
  • 3D Ergo technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Especificaciones

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Blue, Gray
Hand Ambidextrous
Latex Free Yes
Material Dyneema, Polyamide, Elastane
Product Type Cut-resistant Gloves
Size 7
EN388 Standard 4X42C
For Use With (Application) Food Processing, Inspection, Precision Work, and Sorting
Gender Unisex
Glove Cuff Style Knit
Series Phynomic C5
Yarn Gauge 15
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.