missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic C5 Cut Protection Gloves
GREENER_CHOICE

Product Code. 17290391 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17290391 11 10 pairs
17240391 6 10 pairs
17250391 7 10 pairs
17260391 8 10 pairs
17270391 9 10 pairs
17280391 10 10 pairs
17200401 12 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17290391 Supplier uvex Supplier No. 6008111
  • ONLINE EXCLUSIVE PRICE
    159.00 €
    156.80 € / 10 pairs
    Save 2.20 €
    1% Off
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight, sensitive gloves provide level C cut resistance for mechanical activities. Aqua-polymer foam coating is breathable and enhances grip capabilities in dry or slightly damp applications.

  • Lightweight construction acts as a second-skin for tactile sensitivity
  • Optimum material composition provides level C cut protection
  • Aqua-polymer foam coating is breathable and repels dampness
  • Gloves are made from materials that prevent excessive perspiration
  • Enhanced grip in dry, and slightly damp applications
  • 3D Ergo technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Blue, Gray
Hand Ambidextrous
Latex Free Yes
Material Dyneema, Polyamide, Elastane
Product Type Cut-resistant Gloves
Size 11
EN388 Standard 4X42C
For Use With (Application) Food Processing, Inspection, Precision Work, and Sorting
Gender Unisex
Glove Cuff Style Knit
Series Phynomic C5
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.