Learn More
uvex Phynomic FOAM Protective Gloves with Aqua-Polymer Foam Coating
Product Code. 15386902 Shop All Uvex Products
Change view
Click to view available options
Size:
5
8
9
10
12
Unit Size:
10 pairs
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15386902 9 Each
17757579 5 10 pairs
15325612 8 Each
15345612 10 Each
17290361 12 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15386902 Supplier uvex Supplier No. 60050099white/grey
  • ONLINE EXCLUSIVE PRICE
    12.25 €
    5.87 € / Pack of 1
    Save 6.38 €
    52% Off
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
Phynomic series gloves are ideal for all types of work where a high level of feeling is required. These safety gloves act as a second skin and are especially light and flexible, without making hands immediately sweaty. The aqua-polymer foam coating enhances grip capabilities in dry environments.
  • High dexterity safety glove designed for precision mechanical work
  • Damp-resistant, aqua-polymer foam coating provides mechanical abrasion resistance
  • Highly-breathable palm coating enhances grip capabilities in dry and slightly damp areas
  • Tactile feel when assembling small parts
  • Ideal for precision assembly work, inspection, sorting, food processing, and packaging
TRUSTED_SUSTAINABILITY
Specifications
Construction Seamless Knit
Coating Material Aqua-Polymer Foam
Color White, Gray
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Spandex, Polyamide
Product Type Mechanical Protection Gloves
Size 9
Style Full-hand Mitt
EN388 Standard 3121X
Glove Cuff Style Knit
Series Phynomic FOAM