Learn More
uvex Phynomic FOAM Protective Gloves with Aqua-Polymer Foam Coating
Product Code. 17757579 Shop All Uvex Products
Change view
Click to view available options
Size:
5
8
9
10
12
Unit Size:
10 pairs
Each
Product Code. Size unitSize
17757579 5 10 pairs
15325612 8 Each
15345612 10 Each
15386902 9 Each
17290361 12 10 pairs
5 options
Product Code. 17757579
Brand: uvex 6005005
Only null left
Add to basket
Add to basket
Phynomic series gloves are ideal for all types of work where a high level of feeling is required. These safety gloves act as a second skin and are especially light and flexible, without making hands immediately sweaty. The aqua-polymer foam coating enhances grip capabilities in dry environments.
  • High dexterity safety glove designed for precision mechanical work
  • Damp-resistant, aqua-polymer foam coating provides mechanical abrasion resistance
  • Highly-breathable palm coating enhances grip capabilities in dry and slightly damp areas
  • Tactile feel when assembling small parts
  • Ideal for precision assembly work, inspection, sorting, food processing, and packaging
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 388 (3 1 2 1 X)
Construction Seamless Knit
Coating Material Aqua-Polymer Foam
Color White, Gray
Exterior Finish Smooth
Hand Ambidextrous
Material Spandex, Polyamide
Product Type Mechanical Protection Gloves
Size 5
Style Full-hand Mitt
EN388 Standard 3121X
Glove Cuff Style Knit
Series Phynomic FOAM