missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic Lite Gray Safety Gloves

Product Code. 17290341 Shop All Uvex Products
Change view
Click to view available options
Size:
5
6
7
8
9
10
11
12
Unit Size:
10 pairs
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17290341 8 10 pairs
17260341 5 10 pairs
17270341 6 10 pairs
17280341 7 10 pairs
17200351 9 10 pairs
17210351 10 10 pairs
17220351 12 10 pairs
17747579 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17290341 Supplier uvex Supplier No. 6004008
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight gloves are constructed from a combination of polyamide and elastane, with aqua polymer impregnation on the palm and fingertips for increased durability and dexterity.

  • Extremely lightweight construction makes these gloves suitable for particularly delicate tasks
  • Aqua-polymer impregnation on the palm and fingertips for mechanical abrasion resistance
  • Enhances grip capabilities in dry and slightly damp areas
  • Porous coating is breathable to reduce perspiration
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
  • 3D Ergo technology ensures a secure fit
  • Suitable for use with food handling and processing
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer
Color Gray
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Elastane, Polyamide
Product Type Mechanical Protection Gloves
Size 8
EN388 Standard 2121X
For Use With (Application) Driving/Operating Machinery, Painting, and Paper Hanging
Gender Unisex
Glove Cuff Style Knit
Series Phynomic Lite
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.