missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic Pro Safety Gloves

Product Code. 17290371 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17290371 6 10 pairs
17200381 7 10 pairs
17210381 8 10 pairs
17220381 9 10 pairs
17230381 10 10 pairs
17240381 11 10 pairs
17250381 12 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17290371 Supplier uvex Supplier No. 6006206
  • ONLINE EXCLUSIVE PRICE
    73.50 €
    65.78 € / 10 pairs
    Save 7.72 €
    10% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Second skin design is light and flexible to help minimize excessive perspiration, while aqua-polymer foam coating repels dirt and moisture.

  • Lightweight construction acts as a second-skin for tactile sensitivity
  • Bamboo/polyamide/elastane liner provides a high level of user comfort
  • Natural fibers help to absorb water vapor
  • Aqua-polymer foam coating repels dirt and moisture
  • Breathable materials help prevent excessive perspiration
  • Enhanced grip in dry, damp, and oily applications
  • 3D Ergo technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Pro
Color Blue, Gray
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Bamboo, Polyamide, Elastane
Product Type Mechanical Protection Gloves
Size 6
EN388 Standard 2121X
For Use With (Application) Assembly, Construction, Maintenance, Outdoor Activities, and Repairs
Gender Unisex
Glove Cuff Style Knit
Series Phynomic Pro
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.