Learn More
uvex Phynomic WET Water-Resistant Gloves with Aqua-Polymer Foam Coating
Product Code. 15306912 Shop All Uvex Products
Change view
Click to view available options
Size:
8
9
10
11
12
Unit Size:
10 pairs
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15306912 11 Each
15336912 8 Each
15346912 9 Each
15396902 10 Each
17270371 12 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15306912 Supplier uvex Supplier No. 6006011
  • ONLINE EXCLUSIVE PRICE
    6.15 €
    6.07 € / Pack of 1
    Save 0.08 €
    1% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Uvex™ Phynomic series gloves are ideal for all types of work where a high level of feeling is required. These safety gloves act as a second skin and are especially light and flexible, without making hands immediately sweaty. The aqua-polymer foam coating offers a high level of protection against humidity.
  • Water-repellent safety gloves with an aqua-polymer foam coating for use in outdoor areas
  • Breathable coating is durable to provide a high degree of mechanical abrasion resistance
  • Enhanced grip capabilities in damp and wet areas
  • Tactile feel when assembling parts
  • Ideal for precision work, maintenance, and repairs
TRUSTED_SUSTAINABILITY
Specifications
Construction Seamless Knit
Coating Material Aqua-Polymer Foam
Color Blue, Black
Exterior Finish Smooth
Hand Ambidextrous
Material Spandex, Polyamide
Product Type Mechanical Protection Gloves
Size 11
Style Full-hand Mitt
EN388 Standard 3131X
Glove Cuff Style Knit
Series Phynomic WET