Learn More
uvex Phynomic XG Safety Gloves with Aqua-Polymer Xtra Grip Foam Coating
GREENER_CHOICE
Product Code. p-7236908 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15376912 6 Each
15396912 8 Each
15306922 9 Each
15356912 10 Each
15366912 11 Each
15386912 7 Each
17260381 12 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15376912 Supplier uvex Supplier No. 6007006
Add to basket
Edge
Add to basket
Uvex™ Phynomic series gloves are ideal for types of work where a high level of feeling is required. These safety gloves act as a second skin and are especially light and flexible, without making hands immediately sweaty. The Xtra Grip aqua-polymer foam coating enhances grip in oily conditions.
  • Xtra Grip aqua-polymer foam coating is breathable and provides a high level of mechanical abrasion resistance
  • Enhanced grip capabilities in oily areas
  • Tactile feel when assembling oily parts
  • Ideal for precision work, maintenance, repair work, and metal processing
TRUSTED_SUSTAINABILITY
Specifications
Construction Seamless Knit
Coating Material Aqua-Polymer Foam
Color Black, Gray
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Spandex, Polyamide
Product Type Mechanical Protection Gloves
Size 6
Style Full-hand Mitt
EN388 Standard 4121X
Glove Cuff Style Knit
Series Phynomic XG