missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PI Lipid Kinase Substrate

Product Code. 10553274
Change view
Click to view available options
Quantity:
800 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10553274 800 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10553274 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' PV5371

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifically optimized for use with the Class II (PIK3C2A & PIK3C2B) PI3 kinases.

Specifically optimized for use with the Class II (PIK3C2A & PIK3C2B) PI3 kinases. These vesicles consist of 100% phosphatidylinositol (PI).

For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Biochemical ADP accumulation
Chromagen Quantity 800nmol
Content And Storage PI Lipid Kinase Substrate in 50 mM HEPES, pH 7.5

Store at -20°C.
Detection Method Fluorescence
Form Frozen
For Use With (Application) Pharma and biopharma, target and lead identification and validation, drug discovery and development, kinase biology, biochemical kinase assays, Adapta™ universal kinase assays
Quantity 800 nmol
Substrate Phosphatidylinositol (PI) vesicles
Substrate Properties Lipid-Based Substrate, Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Lipid Kinases
Item Description PI Lipid Kinase Substrate, 800nmol, Specifically optimized For Use With the Class II (PIK3C2A & PIK3C2B) PI3 Kinases, These Vesicles Consist of 100% Phosphatidylinositol (PI), For Use With the Adapta™ Universal Kinase Assay (PV5099)
Product Type PI Lipid Kinase Substrate
Shipping Condition Dry Ice
Storage Requirements –20°C
Target Entry PIK3C2B, PIK3C2A
Validated Kinases PIK3C2B
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.