missing translation for 'onlineSavingsMsg'
Learn More

Cytiva pI Markers

Product Code. 10649084 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
2.5 to 6.5
3 to 10
5 to 10.5
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10649084 3 to 10 Each
10024594 2.5 to 6.5 Each
10616515 5 to 10.5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10649084 Supplier Cytiva Supplier No. 17047101
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Run these markers in parallel with the experimental samples on isoelectric focusing gels. Cytiva Life Sciences™ pI Markers are lyophilized mixtures of stable, salt-free, highly purified proteins.

The markers may be run in parallel with the experimental samples on isoelectric focusing gels and are not prestained.

  • The precisely defined pI of the markers allows a permanent and accurate calibration of sample components
  • pH gradients and pI can be measured to within 0.01 to 0.05 pH unit
  • The progress of the focusing experiment can be monitored visually by the focusing of methyl red or Cytochrome c

Not for use in denaturing (urea) systems.

TRUSTED_SUSTAINABILITY

Specifications

Color White
pH Range 3 to 10
Form Lyophilized
For Use With (Application) To be used for isoelectric focusing (IEF)
Kit Contents Amylglucosidase - 3.50 pI (Native), Methyl red (dye) - 3.75 (pI Native), Trypsin inhibitor - 4.55 (pI Native), b-Lactoglobulin A - 5.20 (pI Native), Carbonic Anhydrase B (Bovine) - 5.85 (pI Native), Carbonic Anhydrase B (human) - 6.55 (pI Native)
Quantity 1 Kit: 10 Vials
Range 3.5 to 9.3 pl at 24°C
Storage Requirements 4°C
Product Type pl Marker Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.