missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PI:PS Lipid Kinase Substrate

Product Code. 10786221
Change view
Click to view available options
Quantity:
800 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10786221 800 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10786221 Supplier Thermo Scientific™ Supplier No. PV5122
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifically optimized for use with the Class III PI3 kinase (PIK3C3 (hVPS34)) and Type III PI4 lipid kinases, this substrate can also be used as a general substrate for the Class I lipid kinases.

Specifically optimized for use with the Class III PI3 kinase (PIK3C3 (hVPS34)) and Type III PI4 lipid kinases, this substrate can also be used as a general substrate for the Class I lipid kinases. The lipid vesicles consist of a 1:9 molar ratio of phosphatidylinositol (PI) to phosphoserine (PS).

For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Biochemical ADP accumulation
Content And Storage PI:PS Lipid Kinase Substrate in 50 mM HEPES, pH 7.5.

Store at -20°C.
Detection Method Fluorescence
Form Liquid
For Use With (Application) Pharma and biopharma, protein assays and analysis, target and lead identification and validation
Quantity 800 nmol
Substrate Phosphatidylinositol (PI) lipid vesicles with phosphoserine (PS)
Substrate Properties Lipid-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Lipid Kinases
Gene ID (Entrez) 5289
Item Description PI:PS Lipid Kinase Substrate, 800nmol, 5289 Gene ID, Fluorescent Detection Method, Lipid Kinases Target Enzyme, Used For Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation
Packaging 1 Each
pH 7.5
Product Type PI: PS Lipid Kinase Substrate
Shipping Condition Dry Ice
Storage Requirements –20°C
Target Entry PIK3C3, PI4KA, PI4KB
Validated Kinases PIK3CG, PIK3C3, PI4KB
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.