missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Pi-Pump™ Green Pipette

Product Code. 17200634 Shop All Bürkle Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17200634 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17200634 Supplier Buerkle™ Supplier No. 75510002

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Proven thousands of times, Buerkle™ Pi-Pump™ Green Pipettes fit naturally into the hand.

  • Handwheel ensures precise suction
  • Contents can be purged by pressing back the piston
  • Dispensing with the lateral switch
  • Easy to use
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Product Type Pipette
For Use With (Equipment) Fits Pipette up to 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.