missing translation for 'onlineSavingsMsg'
Learn More

Picolinic Acid Ethyl Ester, TRC

Product Code. 30428556
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428556 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428556 Supplier Toronto Research Chemicals Supplier No. TRCP4372351G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Picolinic Acid Ethyl Ester
CAS 2524-52-9
Molecular Formula C8H9NO2
Quantity 1 g
Synonym 2-Pyridinecarboxylic Acid Ethyl Ester, Ethyl Pyridine-2-carboxylate, 2-(Ethoxycarbonyl)pyridine, Ethyl 2-Picolinate, Ethyl 2-Pyridinecarboxylate, Ethyl Picolinate
InChI Formula InChI=1S/C8H9NO2/c1-2-11-8(10)7-5-3-4-6-9-7/h3-6H,2H2,1H3
SMILES CCOC(=O)c1ccccn1
IUPAC Name ethyl pyridine-2-carboxylate
Molecular Weight (g/mol) 151.16
Formula Weight 151.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.