missing translation for 'onlineSavingsMsg'
Learn More

Picric Acid, Saturated, Approximately 1.2% (w/v), Ricca Chemical

Product Code. 31749547
Change view
Click to view available options
Quantity:
100 mL
Packaging:
500 mL HDPE Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31749547 100 mL 500 mL HDPE Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31749547 Supplier Ricca Chemical Company Supplier No. R5860000100A
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5 , 88-89-1
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) Mixture
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
IUPAC Name 2,4,6-trinitrophenol

Specifications

Melting Point 0.0°C
Boiling Point 100°C
CAS Max % 1.18%, 98.82%
Color Yellow
Appearance Yellow Liquid
Quantity 100 mL
CAS Min % 1.18%, 98.82%
Packaging 500 mL HDPE Bottle
Physical Form Liquid
SMILES OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
Grade Laboratory
Chemical Name or Material Picric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.