missing translation for 'onlineSavingsMsg'
Learn More

Picric Acid Solution R1, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11464719 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Tamper evident
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11464719 100 mL Tamper evident 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11464719 Supplier Reagecon Supplier No. 1065802
  • ONLINE EXCLUSIVE PRICE
    56.25 €
    47.46 € / 100mL
    Save 8.79 €
    15% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured according to Eur. Pharm. Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Picric Acid Solution R1, EP Grade, Reagecon™ is a ready-to-use reagent used to prepare crystalline salts of organic bases for the purpose of identification and characterization and in creatine identification.

Picric acid is used in organic chemistry for the preparation of crystalline salts of organic bases (picrates) for the purpose of identification and characterization. It is also used in the Jaffe reaction to test for creatinine; it forms a colored complex that can be measured using spectroscopy. This solution is prepared by combining 100mL of saturated 10g/L picric acid solution with 0.25mL of strong sodium hydroxide solution.
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise reagents and volumetric solutions

Specifications

CAS 88-89-1, 1310-73-2, 7732-18-5
Color Yellow
Molecular Formula C6H3N3O7
Linear Formula (O2N)3C6H2OH
Synonym picric acid, trinitrophenol, carbazotic acid, picronitric acid, melinite, acide picrique, phenol trinitrate, picral, nitroxanthic acid, pikrinsaeure
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name 2,4,6-trinitrophenol
Molecular Weight (g/mol) 229.104
PubChem CID 6954
ChEBI CHEBI:46149
Formula Weight 229.1g/mol
Grade Eu. Ph. Chapter 4(4.1.1)
Concentration or Composition (by Analyte or Components) 100mL saturated 10g/L picric acid solution + 0.25mL strong sodium hydroxide solution
Quantity 100 mL
Packaging Tamper evident
Physical Form Liquid
Chemical Name or Material 2,4,6-Trinitrophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.