missing translation for 'onlineSavingsMsg'
Learn More

Picropodophyllin, MedChemExpress

Product Code. 16439147
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439147 5 mg 5mg
16432667 50 mg 50mg
16426578 10 mg 10mg
16406786 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16439147 Supplier Medchem Express Supplier No. HY15494
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Picropodophyllin (AXL1717) is a selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor with an IC50 of 1 nM.

Picropodophyllin (AXL1717) is a selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor with an IC50 of 1 nM.

Specifications

Chemical Name or Material Picropodophyllin
CAS 477-47-4
Color White
Purity Grade Notes Research
Molecular Formula C22H22O8
Quantity 5 mg
Synonym AXL1717 Picropodophyllin PPP
Solubility Information DMSO : 50 mg/mL (120.65 mM; Need ultrasonic)
SMILES O=C1OC[C@]2([H])[C@@H](O)C3=C(C=C4OCOC4=C3)[C@@H](C5=CC(OC)=C(OC)C(OC)=C5)[C@@]21[H]
Molecular Weight (g/mol) 414.41
Formula Weight 414.41
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.