missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Picropodophyllotoxin

Product Code. 18772484
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18772484 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18772484 Supplier R&D Systems™ Supplier No. 2956/10
  • ONLINE EXCLUSIVE PRICE
    338.64 €
    303.45 € / 10mg
    Save 35.19 €
    10% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective IGF1R inhibitor

Orally active insulin-like growth factor 1 receptor (IGF1R) inhibitor that exhibits no activity at the insulin receptor, FGFR, PDGFR or EGFR. Inhibits IGF1R autophosphorylation (IC50 ∽ 1 nM), increases the fraction of cells in the G2/M phase and upregulates apoptosis. Exhibits antiproliferative effects in multiple cancer cell lines (IC50 = 0.05 - 15 μM), and has anticancer and antineovascularization activity in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 477-47-4
Quantity 10 mg
Product Line Tocris
Molecular Formula C22H22O8
Formula Weight 414.41
Purity >97%
Molecular Weight (g/mol) 414.41
Chemical Name or Material Picropodophyllotoxin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.