missing translation for 'onlineSavingsMsg'
Learn More

Picrotoxin, 95%

Product Code. 11906191 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11906191 25 g Glass Bottle 25g
10607314 1 g Glass Bottle 1g
10676522 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11906191 Supplier Thermo Scientific Acros Supplier No. 131210250
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124-87-8 | C30H34O13 | 602.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-87-8
Molecular Formula C30H34O13
Molecular Weight (g/mol) 602.58
InChI Key VJKUPQSHOVKBCO-DMRKGALJSA-N
Synonym picrotoxin
PubChem CID 131846146
SMILES CC(=C)C1C2C3C4(C(C1C(=O)O2)(CC5C4(O5)C(=O)O3)O)C.CC12C3C4C(C(C1(CC5C2(O5)C(=O)O3)O)C(=O)O4)C(C)(C)O

Specifications

Melting Point 199.0°C to 202.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Mixture of Picrotoxinin and Picrotin) (HPLC)
Packaging Glass Bottle
Quantity 25 g
Merck Index 15,7527
Specific Rotation − 32.00
Solubility Information Solubility in water: 1g/5mL (100°C). Other solubilities: soluble in boiling alcohol (1gr/3mL),dilute acids,and alkalies; sparingly soluble in ether and ccl4,readily soluble in strong ammonia water and in aq.,solns of naoh.,1gr/13.5 mL in 95% ethan
Formula Weight 602.58
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Picrotoxin
Show More Show Less
compliance-icons
Product Identifier
  • Picrotoxin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.