missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Picus™ 2 Electronic Single Channel Pipette
BIOPHARMA

Product Code. 17878469 Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
10 μL
120 μL
300 μL
1000 μL
5000 μL
10000 μL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17878469 300 μL Each
17818479 10 μL Each
17818469 120 μL Each
17828469 1000 μL Each
17838469 5000 μL Each
17888469 10000 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17878469 Supplier Sartorius Supplier No. LH747061
  • ONLINE EXCLUSIVE PRICE
    1360.00 €
    875.00 € / Pack of 1
    Save 485.00 €
    35% Off
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Picus™ 2 Electronic Single Channel Pipette offers reliable, ergonomic pipetting for all users. It has customizable programs, safety features, and connects to the Sartorius app for automatic adjustments and improved workflow, enhancing lab productivity.

  • This electronic single channel pipette is one of the most user-friendly and simple (yet advanced) pipettes on the market.
  • Ideal tool for daily repetitive liquid handling in laboratories, where accuracy, speed, and ergonomics are all important.
  • Fully electronic design and control allows more consistent, reliable, and reproducible results.
  • The precise electronic pipetting system of Picus™ 2 pipettes enable accurate and precise pipetting results, further reducing variance between users.
  • Be kind to your hand: The single-channel Picus™ 2 Electronic Pipette offers truly effortless pipetting thanks to the uniquely light weight (only 100 grams) and fully electronic experience, including tip ejection.
  • Comfortable handle design and finger hook let the pipette rest lightly in your hand.
  • Connect for the future: The Sartorius pipetting mobile app is your handy companion for sample preparation workflows and pipette management. To stay up to date, use the Pipetting app to update your pipettes with the latest features. Open connectivity enables you to integrate Picus 2 into of an existing system
  • Comfortable to use: The single-channel Picus™ 2 Electronic Pipette is accoladed for its superior ergonomic design. With comfort and ease-of-use in mind, it is one of the most user-friendly pipettes. Sartorius has specifically designed this pipette to reduce strain towards user. With today's demanding workloads, this is more important than ever.
  • Reliable and repeatable pipetting results
  • A wide selection of pipetting modes
  • Customizable program
  • Password protection
  • Calibration reminders
  • Sartorius pipetting mobile app
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips Sartorius Optifit Tips, SafetySpace Tips, Flexibulk Tips
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Orange
Disposable Reusable
For Use With Optifit Tip (300μL), SafetySpace Filter Tip
Material Polycarbonate ABS
Number of Channels 1
Programmability Yes
Sterility Non-sterile
Type Electronic Pipette
Volume (Metric) 300 μL
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.