missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Picus™ NxT Electronic Pipettes, 8 Channels
GREENER_CHOICE

Product Code. 15330635 Shop All Sartorius Products
Click to view available options
Volume (Metric):
0.2 to 10 μL
10 to 300 μL
5 to 120 μL
50 to 1200 μL
Color:
Gray
Orange
Purple
Yellow
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15330635

Brand: Sartorius LH745341

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolitik

Designed to ease user's workload and to protect from repetitive strain injury (RSI)

Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.

  • Password protection for stored programs
  • Very last liquid droplets dispensed by repeated blow-out for maximum recovery
  • Features one-touch operating-button and feather-light electronic tip ejection
  • Includes plate tracker for microwell plate dispensing
  • Electronic piston control and brake for precise pipetting
  • Safe-cone filters to prevent contamination
  • Adjustment wheel for volume setting and navigation
  • Range of pipetting modes for diluting and titrating to serial dispensing
  • Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese
  • Certificate of accredited 3-point calibration
  • RFID-enabled to support asset management
  • Volume range: 0.2 μL to 1200 μL
  • Delivered without USB AC adapter (available separately)

Applications:

PCR and other DNA/RNA techniques, ELISA, Protein analysis and Cell culture

Certificeringer

CE Marked

Tjenester

2 year

TRUSTED_SUSTAINABILITY

Tekniske data

Compatible Tips Yellow Optifit Tips, 0.5-200uL (790201); Yellow SafetySpace Tips, 2-120uL (LHLF790101, 790101F)
Color Yellow
Number of Channels 8
Product Line Picus
Type Electronic Pipette
Volume (Metric) 5 to 120 μL
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.